Ethyl 2-(benzyloxy)-4-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60386-500MG 60386-1G 60386-5G
77336-842EA 394.49 USD
77336-842 77355-090 77379-166
Ethyl 2-(benzyloxy)-4-fluoro-5-methylbenzoate ≥97%
Ethyl 2-(benzyloxy)-4-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₇FO₃
MW: 288.313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759720
CAS Number: 2755718-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR