Ethyl 1-phenyl-1H-pyrazole-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

60376-500MG 60376-1G 60376-5G
77335-972EA 371.83 USD
77335-972 77355-088 77379-164
Ethyl 1-phenyl-1H-pyrazole-5-carboxylate ≥97%
Ethyl 1-phenyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MW: 216.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204861
CAS Number: 115315-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR