1-(4-Chloro-2-isopropoxyphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

60374-500MG 60374-1G 60374-5G
77339-082EA 548.99 USD
77339-082 77355-086 77379-162
1-(4-Chloro-2-isopropoxyphenyl)ethanol ≥97%
1-(4-Chloro-2-isopropoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34759714
CAS Number: 2901098-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR