5-(4-Bromophenoxy)-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60364-500MG 60364-1G 60364-25G 60364-5G
77330-868EA 284.28 USD
77330-868 77355-082 77388-652 77379-158
5-(4-Bromophenoxy)-2-fluorobenzaldehyde ≥97%
5-(4-Bromophenoxy)-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34759708
CAS Number: 2755724-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR