5-(4-Bromophenoxy)-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60361-500MG 60361-1G 60361-5G 60361-25G
77330-862EA 226.6 USD
77330-862 77355-076 77367-070 77388-648
5-(4-Bromophenoxy)-2-fluorobenzoic acid ≥97%
5-(4-Bromophenoxy)-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₈BrFO₃
MW: 311.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759705
CAS Number: 2755718-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR