(4-Chloro-2-isobutoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

60351-500MG 60351-1G 60351-25G 60351-5G
77330-858EA 301.79 USD
77330-858 77355-072 77388-646 77379-148
(4-Chloro-2-isobutoxyphenyl)methanol ≥97%
(4-Chloro-2-isobutoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClO₂
MW: 214.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759701
CAS Number: 1613412-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR