4-Chloro-2-isobutoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60350-500MG 60350-1G 60350-5G 60350-25G
77330-856EA 201.88 USD
77330-856 77355-070 77367-066 77388-644
4-Chloro-2-isobutoxybenzaldehyde ≥97%
4-Chloro-2-isobutoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
MW: 212.673 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30609632
CAS Number: 1602198-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR