Methyl 4-chloro-2-isobutoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60349-500MG 60349-1G 60349-5G
77338-612EA 503.67 USD
77338-612 77355-068 77379-146
Methyl 4-chloro-2-isobutoxybenzoate ≥97%
Methyl 4-chloro-2-isobutoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34759700
CAS Number: 1794391-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR