4-(Difluoromethoxy)-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

60344-500MG 60344-1G 60344-5G 60344-25G
77330-854EA 251.32 USD
77330-854 77355-064 77367-064 77388-642
4-(Difluoromethoxy)-2-fluoropyridine ≥97%
4-(Difluoromethoxy)-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD28789582
CAS Number: 1806335-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR