2-(Benzyloxy)-4-chloro-1,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

60341-1G 60341-5G 60341-25G
77340-764EA 60.77 USD
77340-764 77367-062 77379-142
2-(Benzyloxy)-4-chloro-1,3-difluorobenzene ≥97%
2-(Benzyloxy)-4-chloro-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉ClF₂O
MW: 254.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759698
CAS Number: 2755721-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR