1-Chloro-2,4-difluoro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60340-500MG 60340-1G 60340-5G 60340-25G
77330-852EA 166.86 USD
77330-852 77346-216 77367-060 77388-640
1-Chloro-2,4-difluoro-3-isopropoxybenzene ≥97%
1-Chloro-2,4-difluoro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉ClF₂O
MW: 206.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044125
CAS Number: 1881332-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR