1-Chloro-3-ethoxy-2,4-difluorobenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
60339-500MG
60339-1G
60339-5G
60339-25G
77330-850EA
173.04
USD
77330-850
77346-456
77367-058
77388-638
1-Chloro-3-ethoxy-2,4-difluorobenzene ≥95%
1-Chloro-3-ethoxy-2,4-difluorobenzene
Precaución: for Research Use Only
Formula:
C₈H₇ClF₂O MW: 192.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28750531 CAS Number: 1806074-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...