Ethyl 3-bromo-5-fluoro-2-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

60332-500MG 60332-1G 60332-5G
77337-798EA 444.96 USD
77337-798 77355-060 77379-138
Ethyl 3-bromo-5-fluoro-2-iodobenzoate ≥97%
Ethyl 3-bromo-5-fluoro-2-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrFIO₂
MW: 372.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759694
CAS Number: 2382948-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR