Methyl 3-bromo-5-fluoro-2-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

60331-500MG 60331-1G 60331-5G
77330-848EA 349.17 USD
77330-848 77355-058 77379-136
Methyl 3-bromo-5-fluoro-2-iodobenzoate ≥97%
Methyl 3-bromo-5-fluoro-2-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrFIO₂
Storage Temperature: Ambient
MDL Number: MFCD34759693
CAS Number: 2384011-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR