(3-Bromo-5-fluoro-2-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

60327-500MG 60327-1G 60327-5G
77336-206EA 382.13 USD
77336-206 77355-054 77379-132
(3-Bromo-5-fluoro-2-methylphenyl)methanol ≥95%
(3-Bromo-5-fluoro-2-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD19440367
CAS Number: 1255208-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR