Ethyl 3,4-dibromothiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

60319-500MG 60319-1G 60319-5G 60319-25G
77330-844EA 128.75 USD
77330-844 77344-264 77367-054 77379-130
Ethyl 3,4-dibromothiophene-2-carboxylate ≥97%
Ethyl 3,4-dibromothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆Br₂O₂S
MW: 313.994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22421459
CAS Number: 153562-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR