Ethyl 3,5-difluoro-4-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

60318-500MG 60318-1G 60318-5G 60318-25G
77330-842EA 220.42 USD
77330-842 77355-050 77367-052 77388-634
Ethyl 3,5-difluoro-4-(methylthio)benzoate ≥97%
Ethyl 3,5-difluoro-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34759688
CAS Number: 2118580-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR