2-Ethoxy-4-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60314-500MG 60314-1G 60314-5G
77336-508EA 388.31 USD
77336-508 77355-046 77379-126
2-Ethoxy-4-methoxy-5-methylbenzaldehyde ≥97%
2-Ethoxy-4-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34759687
CAS Number: 2166835-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR