2-Ethoxy-4-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60312-500MG 60312-1G 60312-5G
77338-610EA 503.67 USD
77338-610 77355-044 77379-124
2-Ethoxy-4-methoxy-5-methylbenzoic acid ≥97%
2-Ethoxy-4-methoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD34759686
CAS Number: 2168859-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR