1-(Benzyloxy)-2-bromo-5-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60308-500MG 60308-1G 60308-5G 60308-25G
77330-840EA 281.19 USD
77330-840 77355-040 77367-050 77388-632
1-(Benzyloxy)-2-bromo-5-methoxy-4-methylbenzene ≥97%
1-(Benzyloxy)-2-bromo-5-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34759683
CAS Number: 2764731-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR