1-Bromo-2-isopropoxy-4-methoxy-5-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
60307-500MG
60307-1G
60307-5G
77335-700EA
363.59
USD
77335-700
77355-038
77379-120
1-Bromo-2-isopropoxy-4-methoxy-5-methylbenzene ≥97%
1-Bromo-2-isopropoxy-4-methoxy-5-methylbenzene
Precaución: for Research Use Only
Formula:
C₁₁H₁₅BrO₂ MW: 259.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759682 CAS Number: 2764733-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...