1-Bromo-2-ethoxy-4-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60306-500MG 60306-1G 60306-5G 60306-25G
77330-838EA 265.74 USD
77330-838 77355-036 77367-048 77388-630
1-Bromo-2-ethoxy-4-methoxy-5-methylbenzene ≥97%
1-Bromo-2-ethoxy-4-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759681
CAS Number: 2383473-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR