(5-Chloro-6-ethoxynaphthalen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

60300-500MG 60300-1G 60300-5G
77339-996EA 707.61 USD
77339-996 77355-034 77379-118
(5-Chloro-6-ethoxynaphthalen-2-yl)methanol ≥95%
(5-Chloro-6-ethoxynaphthalen-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34759675
CAS Number: 2764731-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR