Methyl 5-chloro-6-ethoxy-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

60298-500MG 60298-1G 60298-5G
77339-184EA 560.32 USD
77339-184 77355-032 77379-116
Methyl 5-chloro-6-ethoxy-2-naphthoate ≥97%
Methyl 5-chloro-6-ethoxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO₃
MW: 264.704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759673
CAS Number: 2764729-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR