(2,6-Dichloro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60266-500MG 60266-1G 60266-5G 60266-25G
77330-834EA 95.79 USD
77330-834 77342-572 77367-044 77379-112
(2,6-Dichloro-3-methylphenyl)(methyl)sulfane ≥97%
(2,6-Dichloro-3-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂S
MW: 207.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787043
CAS Number: 159329-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR