2,4-Dichloro-3-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Dichloro-2-iodo-4-methylbenzene

60265-500MG 60265-1G 60265-5G 60265-25G
77330-832EA 145.23 USD
77330-832 77345-138 77367-042 77388-626
2,4-Dichloro-3-iodotoluene ≥97%
2,4-Dichloro-3-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂I
MW: 286.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079748
CAS Number: 206559-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR