(3-Bromo-2,4,6-trimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60262-500MG 60262-1G 60262-5G 60262-25G
77330-830EA 118.45 USD
77330-830 77343-568 77367-040 77379-110
(3-Bromo-2,4,6-trimethylphenyl)(methyl)sulfane ≥97%
(3-Bromo-2,4,6-trimethylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrS
Storage Temperature: Ambient
MDL Number: MFCD34759648
CAS Number: 2764728-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR