(5-Fluoro-2-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60258-500MG 60258-1G 60258-5G 60258-25G
77330-826EA 193.64 USD
77330-826 77355-024 77367-036 77388-622
(5-Fluoro-2-iodophenyl)(methyl)sulfane ≥97%
(5-Fluoro-2-iodophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₆FIS
MW: 268.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489276
CAS Number: 1202898-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR