4-Fluoro-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

60253-500MG 60253-1G 60253-5G 60253-25G
77330-822EA 203.94 USD
77330-822 77355-020 77367-032 77388-618
4-Fluoro-2-(methylthio)benzoic acid ≥97%
4-Fluoro-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₂S
Storage Temperature: Ambient
MDL Number: MFCD08444546
CAS Number: 410545-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR