Ethyl 3-formyl-5-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60251-500MG 60251-1G 60251-5G
77330-820EA 311.06 USD
77330-820 77355-018 77379-108
Ethyl 3-formyl-5-isopropylbenzoate ≥97%
Ethyl 3-formyl-5-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₃
MW: 220.264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759645
CAS Number: 2755719-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR