3-Bromo-2,6-dimethoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

60243-500MG 60243-1G 60243-5G
77337-464EA 414.06 USD
77337-464 77355-014 77379-104
3-Bromo-2,6-dimethoxybenzonitrile ≥97%
3-Bromo-2,6-dimethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD27930985
CAS Number: 872857-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR