2-(2-Fluoro-5-iodo-4-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

60240-500MG 60240-1G 60240-5G
77340-370EA 962.02 USD
77340-370 77355-012 77379-102
2-(2-Fluoro-5-iodo-4-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(2-Fluoro-5-iodo-4-(trifluoromethyl)phenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₇F₄IO₂
MW: 362.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759641
CAS Number: 2755723-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR