1-Iodo-2-isopropoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60228-500MG 60228-1G 60228-5G
77337-024EA 405.82 USD
77337-024 77355-010 77379-098
1-Iodo-2-isopropoxy-4-methylbenzene ≥97%
1-Iodo-2-isopropoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃IO
Storage Temperature: Ambient
MDL Number: MFCD29096186
CAS Number: 1369835-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR