4-(Benzyloxy)-2-iodo-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

60223-500MG 60223-1G 60223-5G 60223-25G
77330-810EA 221.45 USD
77330-810 77355-002 77367-022 77388-608
4-(Benzyloxy)-2-iodo-1-methylbenzene ≥95%
4-(Benzyloxy)-2-iodo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃IO
MW: 324.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26517114
CAS Number: 1052647-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR