2-Methoxy-3-methyl-6-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

60216-500MG 60216-1G 60216-5G
77336-752EA 393.46 USD
77336-752 77355-000 77379-092
2-Methoxy-3-methyl-6-(trifluoromethyl)pyridine ≥97%
2-Methoxy-3-methyl-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD34759633
CAS Number: 2764731-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR