1-Bromo-3-(tert-butyl)-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60199-500MG 60199-1G 60199-5G
77340-278EA 875.5 USD
77340-278 77354-996 77379-088
1-Bromo-3-(tert-butyl)-2-methoxybenzene ≥97%
1-Bromo-3-(tert-butyl)-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233138
CAS Number: 1007375-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR