2-Fluoro-5-iodo-3-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60197-500MG 60197-1G
77340-446EA 1093.86 USD
77340-446 77354-994
2-Fluoro-5-iodo-3-isopropoxybenzaldehyde ≥97%
2-Fluoro-5-iodo-3-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀FIO₂
MW: 308.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759628
CAS Number: 2901105-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR