4-Fluoro-3-formyl-5-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60195-500MG 60195-1G 60195-5G
77339-822EA 651.99 USD
77339-822 77354-992 77379-086
4-Fluoro-3-formyl-5-isopropoxybenzoic acid ≥97%
4-Fluoro-3-formyl-5-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₄
MW: 226.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759626
CAS Number: 2901106-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR