(3-Ethyl-2-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

60178-500MG 60178-1G 60178-5G
77335-586EA 358.44 USD
77335-586 77354-986 77379-082
(3-Ethyl-2-methoxyphenyl)boronic acid ≥97%
(3-Ethyl-2-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414933
CAS Number: 2135736-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR