1,3-Dibromo-4-chloro-2-fluoro-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

60172-500MG 60172-1G 60172-5G 60172-25G
77330-804EA 227.63 USD
77330-804 77354-984 77367-016 77388-606
1,3-Dibromo-4-chloro-2-fluoro-5-methylbenzene ≥95%
1,3-Dibromo-4-chloro-2-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₂ClF
Storage Temperature: Ambient
MDL Number: MFCD11845919
CAS Number: 1160574-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR