1,3-Dibromo-2-fluoro-4-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60171-500MG 60171-1G 60171-5G 60171-25G
77330-802EA 145.23 USD
77330-802 77345-100 77367-014 77388-604
1,3-Dibromo-2-fluoro-4-iodo-5-methylbenzene ≥97%
1,3-Dibromo-2-fluoro-4-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₂FI
Storage Temperature: Ambient
MDL Number: MFCD34759620
CAS Number: 2764731-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR