2,5-Difluoro-4-iodo-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

60165-500MG 60165-1G 60165-5G 60165-25G
77330-800EA 187.46 USD
77330-800 77354-980 77367-012 77388-602
2,5-Difluoro-4-iodo-1,1'-biphenyl ≥97%
2,5-Difluoro-4-iodo-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇F₂I
MW: 316.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759618
CAS Number: 2757730-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR