4,4'-Dimethyl-[1,1'-biphenyl]-3-carbaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
60163-500MG
60163-1G
60163-5G
77335-996EA
372.86
USD
77335-996
77354-976
77379-072
4,4'-Dimethyl-[1,1'-biphenyl]-3-carbaldehyde ≥95%
4,4'-Dimethyl-[1,1'-biphenyl]-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O MW: 210.271 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759616 CAS Number: 1888790-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...