(4-Iodo-1-methyl-1H-pyrazol-5-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

60156-500MG 60156-1G 60156-5G
77330-798EA 310.03 USD
77330-798 77354-972 77379-068
(4-Iodo-1-methyl-1H-pyrazol-5-yl)methanol ≥97%
(4-Iodo-1-methyl-1H-pyrazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₇IN₂O
Storage Temperature: Ambient
MDL Number: MFCD27944039
CAS Number: 1346818-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR