2-(Benzyloxy)-1-iodo-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

60138-500MG 60138-1G 60138-5G
77338-568EA 498.52 USD
77338-568 77354-970 77379-066
2-(Benzyloxy)-1-iodo-3-methoxybenzene ≥95%
2-(Benzyloxy)-1-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃IO₂
Storage Temperature: Ambient
MDL Number: MFCD34759607
CAS Number: 914604-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR