(2-Isopropoxy-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

60134-500MG 60134-1G 60134-5G
77340-226EA 845.63 USD
77340-226 77354-966 77379-062
(2-Isopropoxy-3-methoxyphenyl)methanol ≥97%
(2-Isopropoxy-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD08442532
CAS Number: 99861-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR