2-Isopropoxy-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60133-500MG 60133-1G 60133-5G 60133-25G
77330-796EA 154.5 USD
77330-796 77345-552 77367-010 77388-600
2-Isopropoxy-3-methoxybenzaldehyde ≥97%
2-Isopropoxy-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02629693
CAS Number: 75792-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR