(4-Iodo-2-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60122-500MG 60122-1G 60122-5G 60122-25G
77330-794EA 204.97 USD
77330-794 77354-962 77367-008 77388-598
(4-Iodo-2-methoxyphenyl)(methyl)sulfane ≥97%
(4-Iodo-2-methoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₉IOS
MW: 280.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759603
CAS Number: 2764732-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR