Ethyl 3-methoxy-4-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

60119-500MG 60119-1G 60119-5G
77337-526EA 416.12 USD
77337-526 77354-958 77379-058
Ethyl 3-methoxy-4-(methylthio)benzoate ≥97%
Ethyl 3-methoxy-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃S
Storage Temperature: Ambient
MDL Number: MFCD18905036
CAS Number: 1824282-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR