1,2-Dichloro-3-fluoro-4-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60098-500MG 60098-1G 60098-5G 60098-25G
77330-786EA 228.66 USD
77330-786 77354-952 77367-000 77388-590
1,2-Dichloro-3-fluoro-4-(methylsulfonyl)benzene ≥97%
1,2-Dichloro-3-fluoro-4-(methylsulphonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO₂S
Storage Temperature: Ambient
MDL Number: MFCD34759599
CAS Number: 2764733-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR